Learn More
Ansell HyFlex™ 11-953 Rough Textured Nitrile-Coated Gloves
Product Code. 16019721 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
Product Code. 16019721
Brand: Ansell 11953060
Only null left
Add to basket
Add to basket
Rough texture on the palm protects against oil, while also enhancing grip capabilities in food-safe environments.
  • Rough nitrile palm coating
  • Protects against oil
  • Enhances grip in food environments
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Gray
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Polyamide
Product Type Mechanical Protection Gloves
Size 6
EN388 Standard 4121A
For Use With (Application) Assembly, Maintenance, Metal Fabrication, and General Handling
Glove Cuff Style Knit
Series 11-953
Yarn Gauge 13