Learn More
Ansell HyFlex™ 11-754 - Ultralight Cut-Resistant Gloves
Click to view available options
Size:
6
7
8
9
10
11
12
Description
- Heightened defenses: Developed with INTERCEPT™ Cut Resistance Technology, and satisfying ANSI A4 and EN ISO D cut resistance standards, HyFlex™ 11-754 industrial gloves guard against lacerations and prevent common workplace injuries
- Enhanced tactile sensitivity: Palm-dipped, with nylon liner, these PU-coated gloves benefit from advanced tactile sensitivity and touchscreen compatibility
- Elevated comfort: Ultra-lightweight and fitted to natural hand contours, HyFlex™ 11-754 PPE gloves elicit minimal hand fatigue
- Improved dexterity: These polyurethane gloves' ultralight design also increases flexibility and everyday ease of use
- Certified skin-friendliness: HyFlex™ 11-754 industrial working gloves are Dermatest™-approved for skin-friendliness
Specifications
Certifications/Compliance (Europe) | Dermatologically Tested|Ecovadis label|EN 388:2016 +A1:2018|EN ISO 21420:2020 |
Coated Area | Palm |
Construction | Knit |
Coating Material | Polyurethane |
Color | Gray |
Cut Resistance Rating | ANSI A4 |
Hand | Ambidextrous |
Latex Free | No |
Material | Nylon, Spandex, Polyurethane |
Product Type | Cut- and Abrasion-resistant Glove |