Learn More
Ansell HyFlex™ 11-730 Polyurethane-Coated Gloves with Intercept™ Technology
Product Code. p-7215006 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 16659083
Brand: Ansell 11730060
Only null left
Add to basket
Add to basket
Gloves combine comfort and dexterity with Intercept™ Technology to provide a high level of cut resistance. Polyurethane coating enhances grip capabilities in dry, as well as slightly oily environments.
  • A combination of comfort, sure handling, and cut resistance
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Ideal for safe use in controlling, and handling/assembly applications that involve contact with sharp pieces
  • Designed for use in dry as well as slightly oily environments
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 407:2004, CE 0493, EN 420:2003 + A1:2009
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Cut- and Abrasion-resistant Glove
Size 6
EN388 Standard 4X43D
For Use With (Application) Metalwork, Parts Assembly, and Press Injection Molding
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Lining Lined
Puncture Rating ANSI/ISEA 105-16 Level 3
Series 11-730
Thumb Style Wing
Yarn Gauge 13