Learn More
Ansell HyFlex™ 11-725 Polyurethane-Coated Cut-Resistant Lycra™ Gloves
Product Code. 16649073 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. Size unitSize
16649073 7 12 pairs
16639073 6 12 pairs
16659073 8 12 pairs
16669073 9 12 pairs
16679073 10 12 pairs
16689073 11 12 pairs
6 options
Product Code. 16649073
Brand: Ansell 11725070
Only null left
Add to basket
Add to basket
Gloves combine comfort and dexterity with a high degree of cut resistance for applications that involve sharp objects.
  • A balance of comfort, dexterity, and sure handling
  • High level of cut resistance for safe use in applications involving contact with sharp piece
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Lycra™ in the liner/coating provides enhances dexterity and fit
  • Designed for use in dry, as well as slightly oily environments
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free No
Material Intercept Technology Yarn
Product Type Mechanical Protection Gloves
Size 7
EN388 Standard 4342B
For Use With (Application) Automotive Assembly, Construction, Maintenance, and Metal Fabrication
Glove Cuff Style Knit
Series 11-725
Yarn Gauge 15