Learn More
Ansell Edmont™ HyFlex™ 11-630 Series Gray Polyurethane Medium Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Description
- The Lycra™ in the liners of the gloves as well as the polyurethane coating provides excellent flexibility and fit.
- The specific formulation of the polyurethane dipping provides a secure grip on dry or slightly oily pieces.
- The PU formulation also increases abrasion resistance on the gloves’ work surface.
- With strong cut resistance
- Knitted construction
Specifications
Specifications
| Back Material | Polyurethane |
| Certifications/Compliance | CE Mark, Standard CATEGORY II EN 388 : 4443 |
| Cuff Material | Polyurethane |
| Product Type | Medium Weight Gloves |
| Size | 9 |
| Glove Cuff Style | Knitwrist |
| Liner Material | Dupont™ Kevlar™ aramid fibers |
| Lining | Nylon/Lycra™/Dyneema™/Cotton/Dupont™ Kevlar™ aramid fiber |
| Palm Type | Coated-palm |
| Quantity | 144 Pairs |
| Show More |