Description
- Designed for a wide range of lightweight applications in dry or slightly oily environments
- Ideal for workers who need a higher level of fit, touch and precision than that offered by their current gloves
- Also ideal for applications where cotton gloves or no gloves at all are used despite the risk of scrapes, blisters and minor injuries
- Ideal for final assembly, airco, electrical engines, wire harness assembly, adjusters work
Specifications
Specifications
| Back Material | Polyurethane |
| Certifications/Compliance | CE Mark, EN 388 : 2121 |
| Cuff Material | Polyurethane |
| Coating Material | Polyurethane |
| Product Type | Lightweight Gloves |
| Size | 10 |
| Style | 11-619 |
| Glove Cuff Style | Knit |
| Lining | Nylon |
| Palm Type | Coated-palm |
| Show More |