missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ HyFlex™ 11-280 Sleeves
GREENER_CHOICE

Product Code. 17637455
Change view
Click to view available options
Size:
Narrow
Wide
Unit Size:
Pack of 50
Product Code. Size unitSize
17637455 Wide Pack of 50
17627455 Narrow Pack of 50
17647455 Narrow Pack of 50
17657455 Wide Pack of 50
17667455 Narrow Pack of 50
17677455 Wide Pack of 50
6 options
This item is not returnable. View return policy

Product Code. 17637455

Brand: Ansell™ 11280120W

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ansell™ HyFlex™ 11-280 Sleeves feature a seamless design and options of lengths and widths to ensure an excellent fit for all-day comfort

  • INTERCEPT™ Cut Resistance Technology provides best-in-class cut protection and a soft and cool feel to the worker
  • Engineered to perform in combination with any HyFlex cut resistant glove
  • Available in three versions: Extra Wide, Wide, Narrow
TRUSTED_SUSTAINABILITY

Specifications

Size Wide
Construction Knitted
Product Type Sleeve
Material HPPE Yarn
Color Gray
Sterility Non-sterile
Style Ambidextrous
Disposable Reusable
Closure Type Elastic Upper with Knit Cuff
Seam Type Seamless
Latex Free Yes
Model HyFlex™11-280
For Use With (Application) Glass repair and replacement; Handling metal sheets and panels
Certifications/Compliance REACH Compliant
Certifications/Compliance (Europe) EN420:2003 + A1:2009; Category ll
Length (Metric) 30 cm
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.