Learn More
Hydroxymethyl-N-methylpiperidine, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Hydroxymethyl-N-methylpiperidine |
| CAS | 7583-53-1 |
| Molecular Formula | C7H15NO |
| Quantity | 100 g |
| Synonym | (1-Methylpiperidin-3-yl)methanol, 1-Methyl-3-piperidinemethanol, 3-(Hydroxymethyl)-1-methylpiperidine, N-Methyl-3-hydroxymethylpiperidine, N-Methylpiperidine-3-methanol, NSC 66541 |
| InChI Formula | InChI=1S/C7H15NO/c1-8-4-2-3-7(5-8)6-9/h7,9H,2-6H2,1H3 |
| SMILES | CN1CCCC(CO)C1 |
| IUPAC Name | (1-methylpiperidin-3-yl)methanol |
| Molecular Weight (g/mol) | 129.2 |
| Formula Weight | 129.1154 |