Learn More
Hydroxyfasudil hydrochloride, MedChemExpress
Product Code. 16485908
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16485908
Brand: Medchem Express HY13911A
MedChemExpress Hydroxyfasudil hydrochloride is a ROCK inhibitor, with IC50s of 0.73 and 0.72 μM for ROCK1 and ROCK2, respectively.
Hydroxyfasudil hydrochloride is a ROCK inhibitor, with IC50s of 0.73 and 0.72 μM for ROCK1 and ROCK2, respectively.
Specifications
Chemical Name or Material Hydroxyfasudil hydrochloride
CAS 155558-32-0
Color White
Purity Grade Notes Research
Molecular Formula C14H18ClN3O3S
Quantity 10 mg
Synonym HA-1100 hydrochloride HA 1100 hydrochloride HA1100 hydrochloride
Solubility Information DMSO : 30 mg/mL (87.25 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (9.69 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1NC=CC2=C1C=CC=C2S(=O)(N3CCNCCC3)=O.[H]Cl
Molecular Weight (g/mol) 343.83
Formula Weight 343.83
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.