missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy naphthol blue disodium salt, For ACS analysis, MP Biomedicals™

Product Code. p-4404935 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 11422852

Brand: MP Biomedicals 0215252910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 165660-27-5
Molecular Formula C20H14N2Na2O11S3
Molecular Weight (g/mol) 600.495
InChI Key LXJHFXHZWYBBKJ-LUUCHEBKSA-N
Synonym `-(2-naphtholazo-3, 6-disulfonic acid)-2-naphthol-4-sulfonic acid trisodium salt, calcium indicator on sodium chloride
PubChem CID 131849366
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Formula Weight 598.5
Color Black to Blue
Quantity 10 g
Physical Form Powder
Chemical Name or Material Hydroxy naphthol blue disodium salt, For ACS analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.