Learn More
Macherey-Nagel™ Hydrophobic Phase Separation Papers, Round, MN 617 WA
Product Code. 10554981 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric):
150 mm
Unit Size:
Pack of 100
Product Code. Diameter (Metric) unitSize
10554981 150 mm Pack of 100
1 options
Product Code. 10554981
Brand: Macherey-Nagel™ 430015
Only null left
Add to basket
Add to basket
Suitable for separating water from organic solvents via filtration.
  • Hydrophobic by impregnation with a silicone
  • Can be used to separate water from water-immiscible organic solvents via filtration
  • Fast filtration, smooth surface
Specifications
Basis Weight 85 g/m2
Format Disc
Filtration Speed Fast
For Use With (Application) Separating water from water-immiscible organic solvents
Grade MN 617 WA
Quantity 100/Pk.
Thickness (Metric) 0.2 mm
Diameter (Metric) 150 mm