missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide solution, Honeywell™

Product Code. 16655753 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
4 x 5 L
6 x 1 L
Unit Size:
1 L
5 L
6 L
Pack of 4
Product Code. Quantity unitSize
16655753 5 L 5 L
16635753 1 L 1 L
15954770 6 x 1 L 6 L
16645753 4 x 5 L Pack of 4
4 options
This item is not returnable. View return policy

Product Code. 16655753

Brand: Honeywell Research Chemicals 18304H5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Meets analytical specification of Ph. Nord., 34.5-36.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5 , 7722-84-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name water
SMILES O

Specifications

CAS Min % ≥50.00%, ≥30.00%
CAS Max % <70.00%, <50.00%
Packaging Plastic Bottle
Quantity 5 L
Physical Form Liquid
Grade Reagent pharmacopeia chemistry reagents
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.