Learn More
Thermo Scientific Alfa Aesar HYDROFLUORIC ACID ACS 48% 4X500G
Brand:  Thermo Scientific Alfa Aesar 033258.57
HYDROFLUORIC ACID ACS 48% 4X500G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11327568
  • POA / 2000g
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Hydrofluoric acid, 48-51%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen fluoride