missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid 48%, Supelco™

Product Code. 15886840
Change view
Click to view available options
Quantity:
500 mL
1 L
5 L
Unit Size:
1 L
5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15886840 5 L 5 L
15833870 500 mL 500mL
15843870 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15886840 Supplier Supelco Supplier No. 1003345000
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,ISO,Reag. Ph Eur

Hydrofluoric acid (EMSURE™ grade product) is used for regulated analyses and highly demanding lab applications. It also offers the highest quality and unmatched scope of specifications to give complete control of test conditions and eliminate uncertainties. EMSURE™ grade products comply with all international regulations and can be used in various applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3

Specifications

Melting Point -35°C
Density 1.15 g/mL at 25°C,1.16 g/cm3 at 20°C
Quantity 5 L
Molecular Formula HF
MDL Number MFCD00011346
Synonym Hydrofluoric acid, Hydrofluoric acid solution, Fluoric acid, HF, Hydrofluoric acid, HF
Molecular Weight (g/mol) 20.01
Grade Ungraded
Chemical Name or Material Hydrofluoric Acid 48%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.