Learn More
Hydrofluoric Acid, 40.0%, Suprapur™, MilliporeSigma™
Product Code. 15896840
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896840 2.5 L 2.5 L
15853870 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15896840 Supplier Supelco Supplier No. 1003352500
 Restricted Item
Estimated Shipment: 18-11-2026
Chemical Identifiers
CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym Hydrofluoric acid solution, Fluoric acid
IUPAC Name hydrogen fluoride
SMILES F
Specifications
Melting Point −44°C
Boiling Point 112°C (1013 hPa)
Density 1.13 g/cm3 (at 20°C)
Packaging Plastic Bottle
Assay Percent Range ≥ 40%
Quantity 2.5 L