Learn More
Hydrochloric Acid, 5%, Chem Lab™
Product Code. 11919187
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11919187 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11919187 Supplier Chem-Lab Supplier No. CL02.0318.1000
Specifications
Appearance Colorless
pH <1
Composition Hydrochloric acid 5 weight % solution
Concentration 51g HCl/l H2O
Density 1.02g/mL
Odor Odorless
Quantity 1 L
Physical Form Liquid
Solubility Information Soluble in Water
compliance-icons
Product Identifier
  • Hydrochloric acid 5 weight % solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid