Learn More
Hydrochloric Acid, 1M, Chem Lab™
Product Code. 10621183
Change view
Click to view available options
Quantity:
5 L
10 L
20 L
Unit Size:
10 L
20 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10621183 5 L 5 L
11913133 10 L 10 L
11933133 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10621183 Supplier Chem-Lab Supplier No. CL05.0311.5500
  • ONLINE EXCLUSIVE PRICE
    602.00 NOK
    470.64 NOK / 5 L
    Save 131.36 NOK
    21% Off
 Restricted Item
In Stock
Add to basket
Add to basket
For laboratory use, Titrations, ASTM D-4047
Specifications
Appearance Colorless
pH <1
Composition Hydrochloric acid 1mol/L
Concentration 36.461g HCl/l H2O = 1 N (±0.002/20°C)
Density 1.02g/mL
Odor Odorless
Quantity 5 L
Physical Form Liquid
Solubility Information Soluble in Water
compliance-icons
Product Identifier
  • Hydrochloric acid 1 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid 1