Learn More
Emma Safety Footwear Hydro-Tec™ Comfort Inlay Sole
Product Code. 13224554
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Each
Product Code. Size unitSize
13224554 42 Each
10757145 41 1 pair
10154214 43 1 pair
10236084 46 1 pair
10011954 44 1 pair
10747145 39 1 pair
10001964 37 1 pair
13284544 38 Each
10327674 40 1 pair
10387574 45 1 pair
10576364 48 1 pair
10433424 36 1 pair
10246084 47 1 pair
10525024 35 1 pair
14 options
Product Code. 13224554
Brand: Emma Safety Footwear MM00107042
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Anatomically shaped and interchangeable.
  • Anti-static (ESD) and anti-bacterial
  • Breathable, regulates and absorbs moisture
  • Washable and resilient
Specifications
Color Gray
Size 42
Style Interchangeable