Learn More
Emma Safety Footwear Hydro-Tec™ Business Inlay Soles
Product Code. 15276450
Change view
Click to view available options
Grade:
Anti-static (ESD) and anti-bacterial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Grade Unit Size
15276450 Anti-static (ESD) and anti-bacterial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15276450 Supplier Emma Safety Footwear Supplier No. MM00107145
  • ONLINE EXCLUSIVE PRICE
    173.33 NOK
    119.62 NOK / Pack of 1
    Save 53.71 NOK
    30% Off
Estimated Shipment: 21-12-2026
With high-performance top layer that provides extra comfort.
  • Interchangeable insole
  • Anti-static (ESD) and anti-bacterial
  • Absorbent and resilient
Specifications
Color Beige
Grade Anti-static (ESD) and anti-bacterial
Style Interchangeable
Product Type Business Inlay Soles