Learn More
Hydrazine Hydrate-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Hydrazine Hydrate-d6 |
| CAS | 102096-80-0 |
| Molecular Formula | D4 N2 . D2 O |
| Quantity | 1 g |
| Synonym | Hydrazine-d4, mono(hydrate-d2) (9CI), Water-d2, compd. with hydrazine-d4 (1:1) (9CI), Hydrazine Hydrate-d6 |
| InChI Formula | InChI=1S/H4N2.H2O/c1-2;/h1-2H2;1H2/i/hD6 |
| SMILES | [2H]O[2H].[2H]N([2H])N([2H])[2H] |
| Molecular Weight (g/mol) | 56.097 |
| Formula Weight | 56.086 |