Learn More
HYDRANAL™ - Coulomat AG, Reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm, Solstice
Product Code. 15624500
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
6 x 500 mL
6 x 1 L
Unit Size:
1 L
250mL
500mL
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15624500 500 mL 500mL
15614500 250 mL 250mL
15694490 1 L 1 L
15644500 6 x 500 mL Pack of 6
15634500 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15624500 Supplier Honeywell-Hydranal Supplier No. 34836500ML
  • ONLINE EXCLUSIVE PRICE
    2095.00 NOK
    1881.48 NOK / 500mL
    Save 213.52 NOK
    10% Off
View all promotions
 Restricted Item
In Stock
Add to basket
CAS: 67-56-1
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material HYDRANAL™-Coulomat AG
CAS 7446-09-5, 68007-08-9, 111-42-2, 67-56-1, 288-32-4
Packaging Poly-coated Bottle
Quantity 500 mL
Flash Point 14°C (57.2°F)
UN Number UN1230
Grade Coulometry
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap..
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methanol, 2,2'-iminodiethanol; diethanolamine, imidazole, sulphur dioxide, 1H-Imidazole monohydriodi