Learn More
HYDRANAL™-CompoSolver E, Solstice
Product Code. 15624380
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15624380 1 L 1 L
15634380 2.5 L 2.5 L
15654380 6 x 1 L Pack of 6
15644380 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15624380 Supplier Honeywell-Hydranal Supplier No. 347341L
  • ONLINE EXCLUSIVE PRICE
    1280.00 NOK
    1191.92 NOK / 1 L
    Save 88.08 NOK
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Medium for volumetric one-component Karl Fischer titration (ethanol-based)
Advantages of Hydranal one-component reagents:
  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)
Improved Composition
Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material HYDRANAL™-CompoSolver E
CAS 64-17-5, 693-98-1, 7446-09-5
Density 0.80g/mL
Packaging Poly-coated Bottle
Quantity 1 L
Flash Point 12°C (53.6°F)
UN Number UN1170
Grade Laboratory
compliance-icons compliance-icons compliance-icons
Product Identifier
  • hydranal composolver E
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethanol; ethyl alcohol, 2-Methylimidazole, sulphur dioxide