Learn More
HYDRANAL™ - Composite 1, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Product Code. p-7072140 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
Unit Size:
1 L
500mL
Pack of 6
Product Code. 15634490
Brand: Honeywell-Hydranal 34827500ML
Request Bulk or Custom Format
Specifications
Chemical Name or Material HYDRANAL™-Composite 1
CAS 288-32-4, 7446-09-5, 111-90-0
Density 1.11g/mL
Quantity 500 mL
Flash Point 90°C (194°F)
compliance-icons
Product Identifier
  • HYDRANAL-Composite 1
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
Supplemental information
  • MIXTURE LIST-Contains : 2-(2-Ethoxyethoxy)ethanol, imidazole, 2-Methylimidazole, sulphur dioxide, iodine, 1H-Imidazole monoh