Learn More
Ansell™ Hyd-Tuf™ 52-502 Series Brown Nitrile Medium Weight Gloves
Product Code. 12600952 Shop All Ansell Products
Change view
Click to view available options
Size:
9
10
Unit Size:
12 pairs
Product Code. Size unitSize
12600952 10 12 pairs
12610952 9 12 pairs
2 options
Product Code. 12600952
Brand: Ansell™ 52502100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Supersedes the classic leather general purpose glove in medium-duty applications. More flexible, more comfortable, far more hard-wearing. Offers a close, comfortable fit while safely cushioning the hand.
  • Superior resistance to abrasion and good protection against snags, cuts and puncture
  • High performance nitrile impregnation
  • Excellent dry grip.
  • Makes handling quicker and easier.
  •  Effectively repels grease, oil and dirt
  • Safely machine-washable: will not shrink, stiffen or lose its shape.
  • More hygienic and longer lasting – can be safely and economically re-used.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance Standard CATEGORY II EN 388 : 3111
Cuff Material Nitrile
Coating Material Nitrile
Product Type Medium Weight Gloves
Size 10
Style 52-502
Glove Cuff Style Knit
Lining Cotton Jersey
Palm Type Fully coated-palm
Quantity 144 Pairs
Resistant To Superior resistance to abrasion and good protection against snags, cuts and puncture
Series 52-502