Learn More
Cytiva HyClone™ Tetracycline Screened Characterized Fetal Bovine Serum, US Origin
Product Code. 17602075
Click to view available options
Processing:
HI, Irr - 25 to 40 doses
Heat Inactivated
Unit Size:
500mL
Product Code. 17602075
Brand: Cytiva SH30071.03TIH2540
FBS with undetectable levels of tetracycline
Tetracycline Screened FBS is designed for researchers using tetracycline-regulated gene expression systems in cultured cells and is suitable for expression studies in Tet-on/Tet-off systems.
TRUSTED_SUSTAINABILITY
Specifications
Country of Origin United States
Form Liquid
Specialty Type Tetracycline screened
Quantity 500 mL
Processing HI, Irr - 25 to 40 doses
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤ 10 mg/dL