Learn More
Cytiva HyClone™ Sodium Bicarbonate
Product Code. 10001472
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
10001472 100 mL 100mL
1 options
Product Code. 10001472
Brand: Cytiva SH30033.01
Specifications
Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Content And Storage 15° to 3°C
Description With 75.0g/Sodium Bicarbonate
Concentration 75 g/L
Sterilization Method 0.1 μm Sterile-filtered
Endotoxin Level =1.0EU/mL
pH 7.9 to 9.4
Quantity 100 mL