Learn More
Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media
Product Code. p-7058040
Change view
Click to view available options
Quantity:
1000 mL
5 L
10 L
6 x 1000 mL
With Additives:
Glutamine, Poloxamer 188, Sodium Bicarbonate
None
Without Additives:
No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate
No Phenol Red, No HEPES
Product Type:
PF CHO LS
PF-CHO LS
PF-CHO MPS
Unit Size:
10 L
1000 L
1000mL
5 L
50 L
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity With Additives Without Additives Product Type unitSize
11582650 10 L None No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate PF-CHO MPS 1000 L
10494085 10 L None No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate PF-CHO MPS 10 L
11572650 10 L None No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate PF-CHO MPS 50 L
11571911 6 x 1000 mL Glutamine, Poloxamer 188, Sodium Bicarbonate No Phenol Red, No HEPES PF CHO LS Pack of 6
10443295 1000 mL Glutamine, Poloxamer 188, Sodium Bicarbonate No Phenol Red, No HEPES PF-CHO LS 1000mL
10721694 5 L None No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate PF-CHO MPS 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11582650 Supplier Cytiva Supplier No. SH30333.02
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.
  • Designed for high cell yield and recombinant protein production
  • Allow for direct or sequential adaptation
  • Support growth in multiple cell culture systems
  • Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
  • TRUSTED_SUSTAINABILITY
    Specifications
    Form Powder
    Product Type PF-CHO MPS
    Sterility Non-sterile
    With Additives None
    Without Additives No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate
    Quantity 10 L
    Certifications/Compliance cGMP
    Content And Storage 2° to 8°C, away from light