Learn More
Cytiva HyClone™ Newborn Calf Serum (U.S.)
Product Code. p-7058017
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Irradiated, 25 to 40 doses
None
Unit Size:
1 L
1000mL
100mL
500mL
Product Code. Quantity Processing unitSize
11571851 100 mL None 100mL
11556551 1000 mL None 1000mL
11581851 500 mL None 500mL
16291102 500 mL Irradiated, 25 to 40 doses 500mL
16605755 1000 mL Irradiated, 25 to 40 doses 1 L
5 options
Product Code. 11571851
Brand: Cytiva SH30118.02
Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.
  • Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old
TRUSTED_SUSTAINABILITY
Specifications
Description Newborn Calf Serum (U.S.)
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus