Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin
Product Code. 16231112
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Processing:
Heat inactivated
Irradiated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
Unit Size:
1000mL
500mL
Product Code. Quantity Processing unitSize
16231112 1000 mL Irradiated, 25 to 40 doses 1000mL
16201112 500 mL Irradiated, 25 to 40 doses 500mL
12624237 1000 mL Heat inactivated 1000mL
16221112 500 mL Irradiated, 30 to 45 doses 500mL
12802956 500 mL Irradiated 500mL
12339962 500 mL Heat inactivated 500mL
16251112 1000 mL Irradiated, 30 to 45 doses 1000mL
7 options
Product Code. 16231112
Brand: Cytiva SH30401.02IR2540
A cost-effective alternative to Fetal Bovine Serum (FBS).
  • Carefully collected, processed, and filtered in New Zealand
  • Ensured safety against bovine diseases
  • We follow the same standards in processing of our calf serum as with our fetal bovine serum (FBS) to enable offering the highest quality and traceability
  • HyClone Newborn Calf Serum, New Zealand Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Additional Assays Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG
    Certifications/Compliance USDA Regulations
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
    Quantity 1000 mL
    Sterility Sterile
    Sterilization Method 0.1 μm filtration
    Processing Irradiated, 25 to 40 doses
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 25 EU/mL
    Heat Inactivated No
    Mycoplasma Detected Yes
    USDA Safety Testing Included Yes