Learn More
Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum, EMEA Tested
missing translation for 'orderingAttributeHoverText'
Processing:
Irradiated
Non-Irradiated
missing translation for 'unitSize'
1 L
Description
- Contains three to four times as much available iron and transferrin as FBS or equine serum
Specifications
Specifications
| Age | 16 to 22 weeks |
| Product Type | Calf Bovine Serum |
| Quantity | 1000 mL |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Processing | Non-Irradiated |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Hemoglobin Concentration | ≤ 10 mg/dL |
| Iron Supplemented | Yes |
| Product Line | HyClone |
| Test Methods | European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests |