Learn More
Cytiva HyClone™ HyCell™ CHO Media, Powder
Product Code. 17612085
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Product Code. Quantity unitSize
17612085 5 L 5 L
1 options
Product Code. 17612085
Brand: Cytiva SH30948.01
A versatile, animal-derived component-free (ADCF) cell culture media
  • Developed for Chinese Hamster Ovary (CHO) Cell Cultures in bioprocessing applications
  • Regulatory-friendly, chemically defined, and animal-derived component-free media
  • Specifically formulated through the HyClone™ Metabolic Pathway Design process
  • Provides excellent performance, supports high peak cell density, and enhances product yield and quality
  • Media can be used for batch, fed-batch, or perfusion applications in multiple cell culture platforms
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Product Type CHO Medium
Sterility Sterile
Sterilization Method 0.1 μm Sterile-filtered
Product Line HyCell
Quantity 5 L
Certifications/Compliance Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Content And Storage Store at 2°C to 8°C, Away from Light
Description Complete Powder without Poloxamer 188, L-Glutamine, Hypoxanthine, and Thymidine