Learn More
Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ Gamma Irradiated Characterized Fetal Bovine Serum (Australia)
Product Code. 12694197
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Product Code. Quantity unitSize
12694197 1000 mL 1000mL
1 options
Product Code. 12694197
Brand: Cytiva (Formerly GE Healthcare Life Sciences) SH30084.04IR
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Additional Assays Alkaline Phosphate, Lactate Dehydrogenase, SGPT, SGOT, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Glucose, Iron, TIBC, Percent Saturation, Gamma Globulin, IgG, pH, Osmolality
Content And Storage <–1°C
Product Type Fetal Bovine Serum
Quantity 1000 mL
Sterilization Method Sequential Triple-filtered, 100 nm, True Pool Technology
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus