Learn More
Cytiva HyClone™ ES Screened Characterized Fetal Bovine Serum, US Origin
Product Code. p-7214290
Click to view available options
Quantity:
100 mL
500 mL
Heat Inactivated:
No
Yes
Unit Size:
100mL
500mL
Product Code. 17286886
Brand: Cytiva SH30071.02E
Optimized for the culture of murine embryonic stem cells
ES Screened FBS supports the growth of undifferentiated murine embryonic stem cells. The screening includes plating efficiency, colony morphology, and toxicity tests.
TRUSTED_SUSTAINABILITY
Specifications
Country of Origin United States
Form Liquid
Specialty Type ES cell screened
Quantity 100 mL
Sterilization Method 40 nm filtration
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL