Learn More
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Powder
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 L
2 x 5 L
50 L
Unit Size:
10 L
2.5kg
50 L
Pack of 2
Description
Buffer cell culture with HyClone™ Dulbecco's Phosphate Buffered Saline (PBS), Powder.
Specifications
Specifications
| Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
| Content And Storage | 2° to 8° C |
| Form | Powder |
| Sterility | Non-sterile |
| Without Additives | No Calcium, No Magnesium |
| Quantity | 10 L |