Learn More
Cytiva HyClone™ Donor Equine Serum (U.S. origin)
Product Code. 12694207
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated
Irradiated (25 to 40 kGy)
Unit Size:
1 L
100mL
500mL
Product Code. Quantity Processing unitSize
12694207 100 mL Heat Inactivated 100mL
17246896 500 mL Irradiated (25 to 40 kGy) 1 L
12339822 500 mL Irradiated 500mL
3 options
Product Code. 12694207
Brand: Cytiva SH30074.02HI
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care. Complete traceability back to the original source.
  • Donor herds are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration
  • The horses are bled in an enclosed facility that is specially equipped for aseptic collection
  • Coggins tests for equine infectious anemia are routinely performed on each horse
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
  • TRUSTED_SUSTAINABILITY
    Specifications
    Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
    Quantity 100 mL
    Sterilization Method Sequential-filtered, 100 nm
    Processing Heat Inactivated
    Tested For BVDV,Rabies Virus,Reovirus,Cytopathogenic Agents
    Heat Inactivated Yes