missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum, US Origin
Description
- Meets the requirements of most discriminating FBS users
- Triple filtered through 100 nm pore size-rated filters
- Standard endotoxin and hemoglobin specifications
- Subjected to moderate biochemical analyses
Specifications
Specifications
| Certifications/Compliance | Certificate of Suitability (no. R1-CEP 2000-076) |
| Country of Origin | United States |
| Form | Serum |
| Product Type | Fetal Bovine Serum |
| Quantity | 50 mL |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Grade | Characterized |
| Volume (Metric) | 50 mL |
| Endotoxin Concentration | ≤ 25 EU/mL |
| Hemoglobin Concentration | ≤ 25 mg/dL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?