missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum, US Origin
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
Description
- Meets the requirements of most discriminating FBS users
- Triple filtered through 100 nm pore size-rated filters
- Standard endotoxin and hemoglobin specifications
- Subjected to moderate biochemical analyses
Specifications
Specifications
| Certifications/Compliance | Certificate of Suitability (no. R1-CEP 2000-076) |
| Country of Origin | United States |
| Form | Serum |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Grade | Characterized |
| Volume (Metric) | 500 mL |
| Endotoxin Concentration | ≤ 25 EU/mL |
| Hemoglobin Concentration | ≤ 25 mg/dL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction