Learn More
Cytiva HyClone™ Cell Boost™ 7a Supplement
Description
- Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture
- Supplements cell culture with amino acids, vitamins, salts, trace elements, and glucose
- Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production
- Designed to support modern manufacturing processes
- Chemically defined (CD), animal-derived component-free (ADCF), and free of any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
- Intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system
- Formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines
- Manufactured to meet cGMP manufacturing standards and QC specifications
Spécification
Spécification
| Description | Without: L-glutamine, With: Poloxamer 188 |
| Cell Line | CHO |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Sterility | Non-sterile |
| For Use With (Application) | High-Yield Recombinant Protein Production in CHO Fed-Batch Processes |
| Product Line | HyClone |
| Quantity | 0.5 L |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?