Learn More
Cytiva HyClone™ Cell Boost™ 1 Supplement
Product Code. 11526291
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11526291 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11526291 Supplier Cytiva Supplier No. SH30584.02
 Restricted Item
Estimated Shipment: 30-11-2026
Specifications
Description Cell Boost 1 (R05.2)
Classification Chemically-defined, Protein-free, Animal Origin-free
Form Liquid
Product Type Cell Culture Media
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Glucose
Without Additives No Phenol Red, No Sodium Pyruvate, No Glutamine
Quantity 100 g