Learn More
Cytiva HyClone™ CDM4HEK293™ Media
Product Code. 11506341
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11506341 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11506341 Supplier Cytiva Supplier No. SH30858.02
  • ONLINE EXCLUSIVE PRICE
    1217.36 NOK
    1066.00 NOK / 1000mL
    Save 151.36 NOK
    12% Off
 Restricted Item
In Stock
  • Chemically-defined
  • Animal-derived component free (ADCF) cell culture media
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Sterilization Method 0.2 μm Sterile-filtered
With Additives Sodium Bicarbonate, Poloxamer 188
Without Additives No Phenol Red, No Glutamine, No HEPES
Quantity 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description CDM4HEK293 Media