Learn More
Cytiva HyClone™ AdvanceSTEM™ Neural Differentiation Kit and Growth Supplement
Product Code. 10115993
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10115993 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10115993 Supplier Cytiva Supplier No. SH30878.02
  • ONLINE EXCLUSIVE PRICE
    975.00 NOK
    711.83 NOK / 50mL
    Save 263.17 NOK
    26% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-02-2027
Includes
Kit includes a proprietary basal Neural Differentiation Medium to which an aliquot of AdvanceSTEM Growth Supplement (SH3087802) is added at time of use. Basal Neural Differentiation Medium and Growth Supplement should be used together, but they are also available as separate components.
Developed to facilitate the optimal neural differentiation of Adipose-derived Mesenchymal Stem Cells (HAMSC), Bone Marrow Mesenchymal Stem Cells (HBMSC)and Multipotent Cord Blood Unrestricted Somatic Stem Cells (MCBUSSCs)
TRUSTED_SUSTAINABILITY
Specifications
Classification None
Form Liquid
Product Type Stem Cell Differentiation Media
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
For Use With (Application) Neural Differentiation of Adipose-derived Mesenchymal Stem Cells (HAMSC), Bone Marrow Mesenchymal Stem Cells (HBMSC)and Multipotent Cord Blood Unrestricted Somatic Stem Cells (MCBUSSCs)
Quantity 50 mL
Culture Type Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)