Learn More
Cytiva HyClone™ AdvanceSTEM™ Mesenchymal Stem Cell Expansion Kit
Description
Support expansion and maintenance of undifferentiated human mesenchymal stem cells (hMSCs) with HyClone™ AdvanceSTEM™ Mesenchymal Stem Cell Expansion Kit.
Specifications
Specifications
| Cell Type | Undifferentiated Human Mesenchymal Stem Cells |
| Form | Liquid |
| Product Type | Stem Cell Differentiation Media |
| Sterility | Sterile-filtered |
| Quantity | 1000 mL |
| Content And Storage | Store at 2° to 8°C away from light |
| Description | Mesenchymal Stem Cell Basal Medium |