Learn More
Cytiva HyClone™ AdvanceSTEM Cell Culture Media
Product Code. 10474433
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10474433 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10474433 Supplier Cytiva Supplier No. SH30822.01
  • ONLINE EXCLUSIVE PRICE
    1370.00 NOK
    913.43 NOK / 500mL
    Save 456.57 NOK
    33% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Provides optimized conditions for embryonic stem cell expansion and differentiation.
  • ES cell-qualified with Cytiva HyClone ES Screened FBS no prescreening required
  • Mycoplasma free and sterility tested
  • TRUSTED_SUSTAINABILITY
    Specifications
    Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
    Classification None
    Form Liquid
    Product Type Stem Cell Medium
    Sterility Sterile
    Sterilization Method Sterile-filtered
    With Additives Sodium Pyruvate
    Without Additives No Glutamine
    Quantity 500 mL
    Description AdvanceSTEM IMDM4SC without L-glutamine
    Culture Type Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)