Learn More
UVP Hybridization Oven Bottles and Accessories
missing translation for 'orderingAttributeHoverText'
Product Type:
Acrylic Carousel (for upper chamber)
Carousel Kit
Hybridization Bottle (15 cm L)
Hybridization Bottle (30 cm L)
Rocker Tray
Material:
Borosilicate Glass
Stainless Steel
missing translation for 'unitSize'
Each
Specifications
Specifications
| Closure Material | Polypropylene, PTFE |
| For Use With (Application) | DNA, RNA, or protein identification |
| For Use With (Equipment) | UVP hybridization ovens |
| Length (Metric) | 150 or 300 mm bottles |
| Material | Borosilicate Glass |
| Product Type | Acrylic Carousel (for upper chamber) |