missing translation for 'onlineSavingsMsg'
Learn More

HYAL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16859124 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16859124 20 μL 20µL
16849124 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16859124

Brand: Proteintech 196251AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a member of the hyaluronidase family. Hyaluronidases are endoglycosidase enzymes that degrade hyaluro, one of the major glycosaminoglycans of the extracellular matrix. The regulated turnover of hyaluro plays a critical role in many biological processes including cell proliferation, migration and differentiation. The encoded protein may also play an important role in sperm function. This gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression, and the expression of specific transcript variants may be indicative of tumor status. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and some isoforms may lack hyaluronidase activity. This gene overlaps and is on the same strand as N-acetyltransferase 6 (GCN5-related), and some transcripts of each gene share a portion of the first exon.

Specifications

Antigen HYAL3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HYAL3
Gene Accession No. O43820, Q76HM9, Q8VEI3
Gene Alias Hyal 3, HYAL3, Hyaluronidase 3, hyaluronoglucosaminidase 3, LUCA 3, LUCA14, LUCA3, Lung carcinoma protein 3, Minna14
Gene Symbols HYAL3
Host Species Rabbit
Immunogen HYAL3 Fusion Protein Ag8611
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109685, 300993, 8372
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.