Learn More
Dostmann Electronic Humidcheck Moisture Measuring Instrument
Product Code. 11873771
Click to view available options
Dimensions (L x W x H):
130 x 40 x 21 mm
Unit Size:
Each
Product Code. 11873771
Brand: Dostmann Electronic 50200340
Only null left
Add to basket
Add to basket
Rich, reliable, robust instrument for mois­ture meas­ure­ments of wood and construc­tion mater­ials.
  • Humidcheck Moisture Measuring Instrument
  • Instrument for mois­ture meas­ure­ments of wood and construc­tion mater­ials
  • Humidimeter for wood
Specifications
Type Humidcheck Moisture Measuring Instrument
Dimensions (L x W x H) 130 x 40 x 21 mm
Weight (Metric) 100 g
For Use With (Application) For moisture measurements of wood and construction materials