missing translation for 'onlineSavingsMsg'
Learn More

Proteintech HumanKine™ recombinant human IL-15 protein

Product Code. p-7225801 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Product Code. Quantity unitSize
17610097 10 μg 1000µg
17620097 1000 μg 100µg
17630097 100 μg 10µg
3 options
This item is not returnable. View return policy

Product Code. 17610097

Brand: Proteintech HZ13231000UG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

HumanKine™ recombinant protein

IL-15 is a 4-α-helix bundle cytokine playing a pivotal role in stimulation of both innate and adaptive immune cells. It is produced primarily by keratinocytes, skeletal muscle cells, monocytes, and CD4+ T cells. It is a member of the common gamma chain family. The members of the common gamma chain family include the IL-2, IL-4, IL-7, IL-9, and IL-21 and require binding to the common  chain receptor for activation. It can be used in growth and maintenance of T and NK cells. It is also shown to be used in proliferation and functional effect of T cells for adoptive cell therapy. It is a glycosylated protein and HumanKine IL-15 appear as 12.5-25 kDa bands (PMID: 24587813, 26627006, 27849617, 31250350).

Specifications

Accession Number P40933
Formulation 50 mM Acetate pH 4.0 + 150 mM NaCl
Gene ID (Entrez) 3600
Molecular Weight (g/mol) 11 -23 kDa
Quantity 10 μg
Source HEK293
Storage Requirements Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Endotoxin Concentration <1 EU/μg
Specific Reactivity Human
Recombinant Recombinant
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Activity 0.07-0.37 ng/mL EC50
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.