missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ HumanKine™ Recombinant Human EPO Protein

Product Code. 16801356 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Product Code. Quantity unitSize
16801356 1000 μg 1000µg
16821356 10 μg 10µg
16811356 100 μg 100µg
3 options
This item is not returnable. View return policy

Product Code. 16801356

Brand: Proteintech™ HZ11681000UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells

  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity

Specifications

Gene ID (Entrez) 2056
Molecular Weight (g/mol) 36 kDa, monomer, glycosylated
Quantity 1000 μg
Species Human
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Content And Storage Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Activity Typically ≤ 2.5 ng/mL EC50
Expression System Human embryonic kidney 293 (HEK293) human cell expression system
Form Lyophilized
Purity or Quality Grade > 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.