Learn More
Proteintech HumanKine™ recombinant human EGF protein
Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Specifications
Specifications
| Accession Number | P01133 |
| Formulation | 1 x PBS, See Certificate of Analysis for details |
| Gene ID (Entrez) | 1950 |
| Molecular Weight (g/mol) | 7 kDa, monomer, glycosylated |
| Quantity | 10 μg |
| Source | HEK293 |
| Storage Requirements | Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) |
| Endotoxin Concentration | <1.0 EU/μg |
| Specific Reactivity | Human |
| Recombinant | Recombinant |
| Show More |