Learn More
Proteintech HumanKine™ recombinant human DKK-1 protein
Product Code. 17690067 Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Product Code. 17690067
Brand: Proteintech HZ131410UG
HumanKine™ recombinant protein
DKK-1 (Dickkopf WNT signaling pathway inhibitor 1) is a member of the DKK protein family. Other members of this family include DKK-2, DKK-3, and DKK-4. The DKK family proteins share 35-40 % homology between various members. Recombinant human DKK-1 expressed in HEK293 cells is a 35-45kDa glycosylated protein. It is a secreted protein that functions as a negative regulator of Wnt/β-catenin signaling pathway. Wnt/ β-catenin signaling pathway involves binding of the Wnt ligands to the seven-transmembrane receptor, Frizzled, and the coreceptor lipoprotein-related protein 5 and 6 (LRP5/6). DKK1 forms a ternary complex with LRP5/6 and another receptor, Kremen, followed by endocytosis of this complex and removal of LRP5/6 from the cell surface. It has been suggested that DKK-1 permits anterior development by inhibiting Wnt/ β-catenin signaling, which is essential for posterior patterning in vertebrates. The balance between Wnt signaling and DKK-1 inhibition is critical for bone formation and homeostasis and insufficient or excess DKK-1 activity in bone results in increased or decreased bone density, respectively. (PMID: 17143291; 12050670; 10570958)
Specifications
Accession Number O94907
Formulation 1 x PBS, See Certificate of Analysis for details
Gene ID (Entrez) 22943
Molecular Weight (g/mol) 35-38 kDa
Quantity 10 μg
Source HEK293
Storage Requirements Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Endotoxin Concentration <1 EU/μg
Specific Reactivity Human
Recombinant Recombinant
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Activity 7-70 ng/mL EC50
Form Lyophilized
Purity or Quality Grade >95%